Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC28
Entrez Gene ID
23331
Gene Name
tetratricopeptide repeat domain 28
Gene Aliases
TPRBK
Chromosome Location
Chr.22: 27978014 - 28679865 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145418.1 NP_001138890.1 14 - 15 4362 65
XM_005261405.2 XP_005261462.1 14 - 15 4362 65
XM_006724171.3 XP_006724234.1 13 - 14 13140 65
XM_011530018.2 XP_011528320.1 14 - 15 4541 65
XM_017028673.1 XP_016884162.1 14 - 15 4335 65
GenBank mRNA AB028966.3 - 9 - 10 3155 65
AB665284.1 - 14 - 15 4310 65
BC016465.1 - 6 - 7 906 65

View Full Product Details