The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289000.1 | NP_001275929.1 | 8 - 9 | 1036 | 82 |
NM_001289001.1 | NP_001275930.1 | 8 - 9 | 1046 | 82 | |
NM_153609.3 | NP_705837.1 | 8 - 9 | 1115 | 82 | |
XM_011529989.2 | XP_011528291.1 | 5 - 6 | 742 | 82 | |
GenBank mRNA | AJ319876.1 | - | 7 - 8 | 978 | 82 |
AY055384.1 | - | 8 - 9 | 1115 | 82 | |
AY358398.1 | - | 8 - 9 | 1046 | 82 | |
BC039082.1 | - | 8 - 9 | 1118 | 82 | |
CR456446.1 | - | 8 - 9 | 1036 | 82 |