The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289001.1 | NP_001275930.1 | 16 - 17 | 2186 | 89 |
NM_153609.3 | NP_705837.1 | 16 - 17 | 2255 | 89 | |
GenBank mRNA | AJ319876.1 | - | 15 - 16 | 2118 | 89 |
AY055384.1 | - | 16 - 17 | 2255 | 89 | |
AY358398.1 | - | 16 - 17 | 2186 | 89 |