The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270584.1 | NP_001257513.1 | 12 - 13 | 1712 | 73 |
NM_001270585.1 | NP_001257514.1 | 13 - 14 | 1626 | 73 | |
NM_024726.4 | NP_079002.3 | 13 - 14 | 1835 | 73 | |
NR_073043.1 | 13 - 14 | 1838 | 73 | ||
XM_011511864.1 | XP_011510166.1 | 13 - 14 | 1621 | 73 | |
XM_011511865.2 | XP_011510167.1 | 13 - 14 | 1861 | 73 | |
XM_011511866.1 | XP_011510168.1 | 12 - 13 | 1501 | 73 | |
XM_017004960.1 | XP_016860449.1 | 12 - 13 | 1498 | 73 | |
GenBank mRNA | AK026180.1 | - | - | 692 | 73 |
AK225433.1 | - | - | 692 | 73 | |
AK294105.1 | - | 12 - 13 | 1635 | 73 | |
AK295566.1 | - | 13 - 14 | 1623 | 73 | |
BC028699.1 | - | 13 - 14 | 1841 | 73 | |
BC043504.1 | - | 13 - 14 | 1749 | 73 |