The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285460.1 | NP_001272389.1 | 23 - 24 | 2960 | 71 |
NM_001285461.1 | NP_001272390.1 | 12 - 13 | 1620 | 71 | |
NM_001285462.1 | NP_001272391.1 | 12 - 13 | 1671 | 71 | |
NM_006633.3 | NP_006624.2 | 24 - 25 | 3354 | 71 | |
XM_005248410.2 | XP_005248467.1 | 24 - 25 | 3204 | 71 | |
XM_005248414.1 | XP_005248471.1 | 13 - 14 | 1799 | 71 | |
XM_011543108.1 | XP_011541410.1 | 13 - 14 | 1791 | 71 | |
XM_017008960.1 | XP_016864449.1 | 23 - 24 | 3132 | 71 | |
GenBank mRNA | AB208856.1 | - | 13 - 14 | 1803 | 71 |
AK291066.1 | - | 24 - 25 | 3354 | 71 | |
AK302510.1 | - | 23 - 24 | 2960 | 71 | |
AK302981.1 | - | 11 - 12 | 1406 | 71 | |
AK303054.1 | - | 12 - 13 | 1640 | 71 | |
AY351902.1 | - | 12 - 13 | 1596 | 71 | |
U51903.1 | - | 24 - 25 | 3285 | 71 |