Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IQGAP2
Entrez Gene ID
10788
Gene Name
IQ motif containing GTPase activating protein 2
Gene Aliases
-
Chromosome Location
Chr.5: 76403255 - 76708132 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285460.1 NP_001272389.1 27 - 28 3555 91
NM_001285461.1 NP_001272390.1 16 - 17 2215 91
NM_001285462.1 NP_001272391.1 16 - 17 2266 91
NM_006633.3 NP_006624.2 28 - 29 3949 91
XM_005248410.2 XP_005248467.1 28 - 29 3799 91
XM_005248414.1 XP_005248471.1 17 - 18 2394 91
XM_011543108.1 XP_011541410.1 17 - 18 2386 91
XM_017008960.1 XP_016864449.1 27 - 28 3727 91
GenBank mRNA AB208856.1 - 17 - 18 2398 91
AK291066.1 - 28 - 29 3949 91
AK302510.1 - 27 - 28 3555 91
AK302981.1 - 15 - 16 2001 91
AK303054.1 - 16 - 17 2235 91
AY351902.1 - 16 - 17 2191 91
U51903.1 - 28 - 29 3880 91

View Full Product Details