The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153234.4 | NP_694966.3 | 3 - 4 | 627 | 110 |
GenBank mRNA | AK098400.1 | - | 3 - 4 | 683 | 110 |
AK291034.1 | - | 3 - 4 | 610 | 110 | |
AK307885.1 | - | 3 - 4 | 568 | 110 | |
BC036467.2 | - | 3 - 4 | 546 | 110 |