Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZBBX
Entrez Gene ID
79740
Gene Name
zinc finger B-box domain containing
Gene Aliases
-
Chromosome Location
Chr.3: 167178399 - 167380405 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199201.1 NP_001186130.1 1 - 2 52 47
NM_024687.3 NP_078963.2 1 - 2 52 47
XM_011513144.1 XP_011511446.1 1 - 2 52 47
XM_011513145.2 XP_011511447.2 1 - 2 160 47
XM_011513148.2 XP_011511450.2 1 - 2 52 47
XM_011513149.2 XP_011511451.2 1 - 2 52 47
XM_011513151.2 XP_011511453.2 1 - 2 96 47
XM_011513155.1 XP_011511457.1 1 - 2 52 47
XM_011513156.1 XP_011511458.1 1 - 2 52 47
XM_017007186.1 XP_016862675.1 1 - 2 52 47
XM_017007187.1 XP_016862676.1 1 - 2 52 47
XM_017007188.1 XP_016862677.1 1 - 2 52 47
XM_017007189.1 XP_016862678.1 1 - 2 160 47
XM_017007190.1 XP_016862679.1 1 - 2 52 47
GenBank mRNA AK093296.1 - 1 - 2 38 47
DB078376.1 - 1 - 2 52 47

View Full Product Details