The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167903.1 | NP_001161375.1 | 10 - 11 | 1189 | 64 |
NM_001289406.1 | NP_001276335.1 | 10 - 11 | 1193 | 64 | |
NM_001289407.1 | NP_001276336.1 | 11 - 12 | 1275 | 64 | |
NM_022736.2 | NP_073573.2 | 11 - 12 | 1306 | 64 | |
NR_110328.1 | 10 - 11 | 1193 | 64 | ||
XM_006713730.2 | XP_006713793.1 | 10 - 11 | 1124 | 64 | |
GenBank mRNA | AB014732.1 | - | 11 - 12 | 1141 | 64 |
AF351617.2 | - | 11 - 12 | 1002 | 64 | |
AK024215.1 | - | 11 - 12 | 1109 | 64 | |
AK294628.1 | - | 10 - 11 | 1066 | 64 | |
AK297593.1 | - | 11 - 12 | 1306 | 64 | |
AK300497.1 | - | 10 - 11 | 992 | 64 | |
AK301680.1 | - | 11 - 12 | 1045 | 64 | |
BC030542.1 | - | 10 - 11 | 1062 | 64 | |
BC042197.1 | - | 11 - 12 | 1118 | 64 |