The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002293.3 | NP_002284.3 | 24 - 25 | 4370 | 83 |
GenBank mRNA | BC015586.2 | - | 6 - 7 | 727 | 83 |
J03202.1 | - | 24 - 25 | 4372 | 83 | |
X13939.1 | - | 2 - 3 | 271 | 83 |