The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002293.3 | NP_002284.3 | 25 - 26 | 4572 | 85 |
GenBank mRNA | BC015586.2 | - | 7 - 8 | 929 | 85 |
DA401791.1 | - | 2 - 3 | 238 | 85 | |
J03202.1 | - | 25 - 26 | 4574 | 85 | |
M27654.1 | - | 1 - 2 | 139 | 85 | |
X13939.1 | - | 3 - 4 | 473 | 85 |