The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102450.2 | NP_001095920.1 | 2 - 3 | 190 | 95 |
XM_005245555.3 | XP_005245612.1 | 2 - 3 | 204 | 95 | |
XM_011510090.2 | XP_011508392.1 | 5 - 6 | 931 | 95 | |
XM_017002635.1 | XP_016858124.1 | 3 - 4 | 706 | 95 | |
XM_017002636.1 | XP_016858125.1 | 1 - 2 | 188 | 95 | |
XM_017002637.1 | XP_016858126.1 | 2 - 3 | 386 | 95 | |
GenBank mRNA | AK294660.1 | - | 2 - 3 | 157 | 95 |
AK296514.1 | - | 2 - 3 | 150 | 95 | |
BC069677.1 | - | 2 - 3 | 139 | 95 | |
BC103865.1 | - | 2 - 3 | 166 | 95 | |
BC103866.1 | - | 2 - 3 | 166 | 95 | |
BC103867.1 | - | 2 - 3 | 166 | 95 | |
BG195915.1 | - | 2 - 3 | 191 | 95 |