Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGL1
Entrez Gene ID
23179
Gene Name
ral guanine nucleotide dissociation stimulator like 1
Gene Aliases
RGL
Chromosome Location
Chr.1: 183636047 - 183928551 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297669.1 NP_001284598.1 14 - 15 2066 60
NM_001297670.1 NP_001284599.1 13 - 14 1956 60
NM_001297671.1 NP_001284600.1 13 - 14 1692 60
NM_001297672.1 NP_001284601.1 12 - 13 1605 60
NM_015149.4 NP_055964.3 14 - 15 2067 60
XM_011509339.2 XP_011507641.1 15 - 16 2113 60
XM_011509341.1 XP_011507643.1 14 - 15 2020 60
XM_011509342.2 XP_011507644.1 14 - 15 2020 60
XM_017000756.1 XP_016856245.1 13 - 14 1541 60
GenBank mRNA AB023176.1 - 13 - 14 1632 60
AF186779.1 - 13 - 14 1653 60
AF186780.1 - 14 - 15 2029 60
AK295179.1 - 13 - 14 1830 60
AK295611.1 - 10 - 11 1269 60
AK304293.1 - 12 - 13 1530 60
AK316526.1 - 14 - 15 2004 60
AL080117.1 - 12 - 13 1435 60
BC136591.1 - 14 - 15 1771 60
BX537416.1 - 14 - 15 2007 60

View Full Product Details