The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297669.1 | NP_001284598.1 | 5 - 6 | 1022 | 70 |
NM_001297670.1 | NP_001284599.1 | 4 - 5 | 912 | 70 | |
NM_001297671.1 | NP_001284600.1 | 4 - 5 | 648 | 70 | |
NM_001297672.1 | NP_001284601.1 | 4 - 5 | 648 | 70 | |
NM_015149.4 | NP_055964.3 | 5 - 6 | 1023 | 70 | |
XM_011509339.2 | XP_011507641.1 | 6 - 7 | 1069 | 70 | |
XM_011509341.1 | XP_011507643.1 | 5 - 6 | 976 | 70 | |
XM_011509342.2 | XP_011507644.1 | 5 - 6 | 976 | 70 | |
XM_017000756.1 | XP_016856245.1 | 4 - 5 | 497 | 70 | |
GenBank mRNA | AB023176.1 | - | 4 - 5 | 588 | 70 |
AF186779.1 | - | 4 - 5 | 609 | 70 | |
AF186780.1 | - | 5 - 6 | 985 | 70 | |
AK295179.1 | - | 4 - 5 | 786 | 70 | |
AK304293.1 | - | 4 - 5 | 573 | 70 | |
AK311701.1 | - | 4 - 5 | 494 | 70 | |
AK311716.1 | - | 4 - 5 | 648 | 70 | |
AK316526.1 | - | 5 - 6 | 960 | 70 | |
AL080117.1 | - | 3 - 4 | 391 | 70 | |
BC136591.1 | - | 5 - 6 | 727 | 70 | |
BX537416.1 | - | 5 - 6 | 963 | 70 |