Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF21B
Entrez Gene ID
23046
Gene Name
kinesin family member 21B
Gene Aliases
-
Chromosome Location
Chr.1: 200969386 - 201023700 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252100.1 NP_001239029.1 24 - 25 3730 62
NM_001252102.1 NP_001239031.1 24 - 25 3730 62
NM_001252103.1 NP_001239032.1 24 - 25 3730 62
NM_017596.3 NP_060066.2 24 - 25 3730 62
XM_017000731.1 XP_016856220.1 23 - 24 3373 62
XM_017000732.1 XP_016856221.1 18 - 19 2514 62
GenBank mRNA AB007918.2 - 24 - 25 3730 62
BC137281.1 - 24 - 25 3438 62
BC144557.1 - 24 - 25 3438 62

View Full Product Details