Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZCCHC11
Entrez Gene ID
23318
Gene Name
zinc finger CCHC-type containing 11
Gene Aliases
PAPD3, TUT4
Chromosome Location
Chr.1: 52423275 - 52553463 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009881.2 NP_001009881.1 21 - 22 3764 95
NM_015269.2 NP_056084.1 21 - 22 3764 95
XM_005270676.2 XP_005270733.1 21 - 22 3773 95
XM_005270678.2 XP_005270735.1 21 - 22 3773 95
XM_005270679.2 XP_005270736.1 21 - 22 3773 95
XM_005270680.2 XP_005270737.1 21 - 22 3773 95
XM_005270681.2 XP_005270738.1 20 - 21 3659 95
XM_005270682.2 XP_005270739.1 19 - 20 3440 95
XM_005270683.2 XP_005270740.1 17 - 18 3407 95
XM_006710498.2 XP_006710561.1 21 - 22 3710 95
XM_006710499.3 XP_006710562.1 21 - 22 3717 95
XM_006710500.1 XP_006710563.1 20 - 21 3560 95
XM_011541097.1 XP_011539399.1 22 - 23 3795 95
XM_011541098.1 XP_011539400.1 21 - 22 3692 95
XM_011541099.1 XP_011539401.1 20 - 21 3479 95
XM_011541100.2 XP_011539402.1 21 - 22 3773 95
XM_017000803.1 XP_016856292.1 21 - 22 3692 95
XM_017000804.1 XP_016856293.1 21 - 22 3692 95
XM_017000805.1 XP_016856294.1 20 - 21 3659 95
XM_017000806.1 XP_016856295.1 20 - 21 3560 95
XM_017000807.1 XP_016856296.1 20 - 21 3560 95
XM_017000808.1 XP_016856297.1 19 - 20 3440 95
XM_017000809.1 XP_016856298.1 19 - 20 3440 95
XM_017000810.1 XP_016856299.1 19 - 20 3440 95
XM_017000811.1 XP_016856300.1 17 - 18 3407 95
XM_017000812.1 XP_016856301.1 17 - 18 3407 95
XM_017000813.1 XP_016856302.1 17 - 18 3407 95
XM_017000814.1 XP_016856303.1 15 - 16 3074 95
GenBank mRNA AK025698.1 - 5 - 6 418 95
AK307579.1 - 21 - 22 3773 95
BC131734.1 - 21 - 22 3656 95
D83776.1 - 20 - 21 3129 95

View Full Product Details