Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ZCCHC11
- Entrez Gene ID
-
23318
- Gene Name
- zinc finger CCHC-type containing 11
- Gene Aliases
-
PAPD3, TUT4
- Chromosome Location
-
Chr.1: 52423275 - 52553463 on Build GRCh38
View Full Product Details