The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029882.3 | NP_001025053.1 | 4 - 5 | 717 | 112 |
XM_005245848.3 | XP_005245905.1 | 5 - 6 | 952 | 112 | |
XM_005245849.3 | XP_005245906.1 | 5 - 6 | 1057 | 112 | |
XM_005245850.3 | XP_005245907.1 | 6 - 7 | 1077 | 112 | |
XM_005245851.3 | XP_005245908.1 | 5 - 6 | 949 | 112 | |
XM_005245852.3 | XP_005245909.1 | 4 - 5 | 726 | 112 | |
XM_011541255.1 | XP_011539557.1 | 5 - 6 | 631 | 112 | |
XM_011541256.2 | XP_011539558.1 | 5 - 6 | 1071 | 112 | |
XM_011541257.2 | XP_011539559.1 | 4 - 5 | 1826 | 112 | |
GenBank mRNA | BC064935.1 | - | 5 - 6 | 840 | 112 |