Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKAR
Entrez Gene ID
150709
Gene Name
ankyrin and armadillo repeat containing
Gene Aliases
-
Chromosome Location
Chr.2: 189674292 - 189763059 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011510673.2 XP_011508975.1 12 - 13 3204 122
XM_011510675.2 XP_011508977.1 11 - 12 3099 122
XM_011510676.2 XP_011508978.1 12 - 13 3204 122
XM_011510677.2 XP_011508979.1 12 - 13 3204 122
XM_011510680.1 XP_011508982.1 12 - 13 3204 122
XM_011510681.1 XP_011508983.1 12 - 13 3204 122
XM_011510682.2 XP_011508984.1 12 - 13 3204 122
XM_011510685.2 XP_011508987.1 12 - 13 2760 122
XM_011510686.2 XP_011508988.1 12 - 13 3204 122
XM_011510687.2 XP_011508989.1 11 - 12 2568 122
XM_011510688.1 XP_011508990.1 12 - 13 3204 122
XM_011510689.2 XP_011508991.1 12 - 13 3204 122
XM_011510691.2 XP_011508993.1 8 - 9 1390 122
XM_011510692.2 XP_011508994.1 10 - 11 2404 122
XM_011510693.2 XP_011508995.1 9 - 10 2300 122
XM_011510694.2 XP_011508996.1 8 - 9 2119 122
XM_017003413.1 XP_016858902.1 12 - 13 3204 122
XM_017003414.1 XP_016858903.1 12 - 13 3204 122
XM_017003415.1 XP_016858904.1 7 - 8 1899 122
XM_017003416.1 XP_016858905.1 9 - 10 2311 122
XM_017003418.1 XP_016858907.1 7 - 8 2014 122
XM_017003420.1 XP_016858909.1 7 - 8 1917 122
GenBank mRNA BC044907.1 - 11 - 12 2570 122

View Full Product Details