Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKAR
Entrez Gene ID
150709
Gene Name
ankyrin and armadillo repeat containing
Gene Aliases
-
Chromosome Location
Chr.2: 189674292 - 189763059 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144708.3 NP_653309.3 4 - 5 1281 142
XM_011510673.2 XP_011508975.1 4 - 5 1767 142
XM_011510674.1 XP_011508976.1 4 - 5 1767 142
XM_011510675.2 XP_011508977.1 4 - 5 1767 142
XM_011510676.2 XP_011508978.1 4 - 5 1767 142
XM_011510677.2 XP_011508979.1 4 - 5 1767 142
XM_011510679.2 XP_011508981.1 4 - 5 1767 142
XM_011510680.1 XP_011508982.1 4 - 5 1767 142
XM_011510681.1 XP_011508983.1 4 - 5 1767 142
XM_011510682.2 XP_011508984.1 4 - 5 1767 142
XM_011510685.2 XP_011508987.1 4 - 5 1323 142
XM_011510686.2 XP_011508988.1 4 - 5 1767 142
XM_011510687.2 XP_011508989.1 3 - 4 1131 142
XM_011510688.1 XP_011508990.1 4 - 5 1767 142
XM_011510689.2 XP_011508991.1 4 - 5 1767 142
XM_017003413.1 XP_016858902.1 4 - 5 1767 142
XM_017003414.1 XP_016858903.1 4 - 5 1767 142
XM_017003417.1 XP_016858906.1 4 - 5 1767 142
GenBank mRNA AJ549812.1 - 3 - 4 980 142
AK126073.1 - 4 - 5 1281 142
BC044907.1 - 3 - 4 1133 142

View Full Product Details