Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKAR
Entrez Gene ID
150709
Gene Name
ankyrin and armadillo repeat containing
Gene Aliases
-
Chromosome Location
Chr.2: 189674292 - 189763059 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144708.3 NP_653309.3 7 - 8 1791 63
XM_011510673.2 XP_011508975.1 7 - 8 2277 63
XM_011510674.1 XP_011508976.1 7 - 8 2277 63
XM_011510675.2 XP_011508977.1 7 - 8 2277 63
XM_011510676.2 XP_011508978.1 7 - 8 2277 63
XM_011510677.2 XP_011508979.1 7 - 8 2277 63
XM_011510679.2 XP_011508981.1 7 - 8 2277 63
XM_011510680.1 XP_011508982.1 7 - 8 2277 63
XM_011510681.1 XP_011508983.1 7 - 8 2277 63
XM_011510682.2 XP_011508984.1 7 - 8 2277 63
XM_011510685.2 XP_011508987.1 7 - 8 1833 63
XM_011510686.2 XP_011508988.1 7 - 8 2277 63
XM_011510687.2 XP_011508989.1 6 - 7 1641 63
XM_011510688.1 XP_011508990.1 7 - 8 2277 63
XM_011510689.2 XP_011508991.1 7 - 8 2277 63
XM_011510691.2 XP_011508993.1 3 - 4 463 63
XM_011510692.2 XP_011508994.1 5 - 6 1477 63
XM_011510693.2 XP_011508995.1 4 - 5 1373 63
XM_011510694.2 XP_011508996.1 3 - 4 1192 63
XM_017003413.1 XP_016858902.1 7 - 8 2277 63
XM_017003414.1 XP_016858903.1 7 - 8 2277 63
XM_017003416.1 XP_016858905.1 4 - 5 1384 63
XM_017003417.1 XP_016858906.1 7 - 8 2277 63
XM_017003418.1 XP_016858907.1 3 - 4 1192 63
XM_017003419.1 XP_016858908.1 3 - 4 1192 63
GenBank mRNA AJ549812.1 - 6 - 7 1490 63
AK126073.1 - 7 - 8 1791 63
BC037215.1 - 4 - 5 1172 63
BC044907.1 - 6 - 7 1643 63
BC047413.1 - 7 - 8 1090 63

View Full Product Details