The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153259.3 | NP_694991.2 | 2 - 3 | 487 | 95 |
XM_005270719.3 | XP_005270776.1 | 2 - 3 | 264 | 95 | |
XM_006710552.2 | XP_006710615.1 | 3 - 4 | 577 | 95 | |
XM_011541187.2 | XP_011539489.1 | 2 - 3 | 321 | 95 | |
XM_011541188.2 | XP_011539490.1 | 2 - 3 | 604 | 95 | |
XM_017000923.1 | XP_016856412.1 | 2 - 3 | 478 | 95 | |
GenBank mRNA | AK309858.1 | - | 2 - 3 | 478 | 95 |
AY083533.1 | - | 1 - 2 | 153 | 95 | |
BC104891.1 | - | 2 - 3 | 305 | 95 | |
BC104893.1 | - | 2 - 3 | 305 | 95 | |
DR007194.1 | - | 2 - 3 | 503 | 95 |