Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRMD3
Entrez Gene ID
257019
Gene Name
FERM domain containing 3
Gene Aliases
4.1O, EPB41L4O, EPB41LO, P410
Chromosome Location
Chr.9: 83242990 - 83560208 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244961.1 NP_001231890.1 1 - 2 108 89
XM_017014589.1 XP_016870078.1 1 - 2 2923 89
GenBank mRNA BC037253.1 - 1 - 2 108 89

View Full Product Details