The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105251.2 | NP_001098721.1 | 7 - 8 | 3030 | 148 |
NM_001284236.1 | NP_001271165.1 | 7 - 8 | 2956 | 148 | |
NM_014733.4 | NP_055548.3 | 6 - 7 | 2902 | 148 | |
XM_005248632.4 | XP_005248689.1 | 6 - 7 | 3028 | 148 | |
XM_011543753.2 | XP_011542055.1 | 7 - 8 | 2927 | 148 | |
XM_017010091.1 | XP_016865580.1 | 8 - 9 | 3088 | 148 | |
XM_017010092.1 | XP_016865581.1 | 7 - 8 | 3074 | 148 | |
XM_017010093.1 | XP_016865582.1 | 6 - 7 | 1408 | 148 | |
XM_017010094.1 | XP_016865583.1 | 5 - 6 | 1353 | 148 | |
XM_017010095.1 | XP_016865584.1 | 8 - 9 | 2982 | 148 | |
XM_017010096.1 | XP_016865585.1 | 3 - 4 | 430 | 148 | |
GenBank mRNA | AB002303.1 | - | 7 - 8 | 2970 | 148 |
AF434817.1 | - | 5 - 6 | 2722 | 148 | |
AL833087.2 | - | 4 - 5 | 1949 | 148 | |
BX537424.1 | - | 6 - 7 | 2854 | 148 | |
CR933621.1 | - | 7 - 8 | 2905 | 148 |