Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CENPF
Entrez Gene ID
1063
Gene Name
centromere protein F
Gene Aliases
CENF, CILD31, PRO1779, STROMS, hcp-1
Chromosome Location
Chr.1: 214603181 - 214664574 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016343.3 NP_057427.3 9 - 10 1493 110
XM_011509082.2 XP_011507384.1 9 - 10 1501 110
XM_017000086.1 XP_016855575.1 9 - 10 1480 110
GenBank mRNA AK307690.1 - 9 - 10 1476 110
U19769.1 - - 1489 110
U30872.1 - 9 - 10 1391 110

View Full Product Details