The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165974.1 | NP_001159446.1 | 20 - 21 | 2128 | 85 |
NM_144639.2 | NP_653240.1 | 19 - 20 | 1948 | 85 | |
GenBank mRNA | AB075869.1 | - | 20 - 21 | 2108 | 85 |
AK055862.1 | - | 19 - 20 | 1926 | 85 | |
BC115405.1 | - | 19 - 20 | 1926 | 85 | |
BC115406.1 | - | 19 - 20 | 1926 | 85 |