Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEMA7A
Entrez Gene ID
8482
Gene Name
semaphorin 7A (John Milton Hagen blood group)
Gene Aliases
CD108, CDw108, H-SEMA-K1, H-Sema-L, JMH, SEMAK1, SEMAL
Chromosome Location
Chr.15: 74409284 - 74433959 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146029.2 NP_001139501.1 8 - 9 1091 87
NM_001146030.2 NP_001139502.1 9 - 10 1029 87
NM_003612.4 NP_003603.1 9 - 10 1133 87
GenBank mRNA AF030698.1 - 9 - 10 1104 87
AF069493.1 - 9 - 10 1109 87
AF071542.1 - 9 - 10 1092 87
AK001895.1 - 3 - 4 374 87
AK293280.1 - 8 - 9 1087 87
AK312257.1 - 9 - 10 1132 87
AM180445.1 - 9 - 10 1092 87
AM180446.1 - 9 - 10 1092 87
AM180447.1 - 9 - 10 1092 87
AM180448.1 - 9 - 10 1092 87
AM180449.1 - 9 - 10 1092 87
AM180451.1 - 9 - 10 1092 87
BC101643.1 - 9 - 10 1124 87
BC101647.1 - 9 - 10 1124 87

View Full Product Details