Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPOCD1
Entrez Gene ID
90853
Gene Name
SPOC domain containing 1
Gene Aliases
PPP1R146
Chromosome Location
Chr.1: 31790422 - 31816051 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281987.1 NP_001268916.1 15 - 16 3094 56
NM_001281988.1 NP_001268917.1 14 - 15 1672 56
XM_011542413.2 XP_011540715.1 15 - 16 1663 56
XM_011542414.2 XP_011540716.1 14 - 15 1858 56
XM_017002779.1 XP_016858268.1 14 - 15 3000 56
XM_017002780.1 XP_016858269.1 13 - 14 2961 56
XM_017002781.1 XP_016858270.1 13 - 14 2877 56
XM_017002782.1 XP_016858271.1 14 - 15 2232 56
GenBank mRNA AK058077.1 - 5 - 6 674 56
AK091016.1 - 14 - 15 1643 56
BC114430.1 - 15 - 16 3020 56

View Full Product Details