Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS9D1
Entrez Gene ID
9605
Gene Name
VPS9 domain containing 1
Gene Aliases
ATP-BL, C16orf7
Chromosome Location
Chr.16: 89707133 - 89721046 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004913.2 NP_004904.2 1 - 2 228 96
XM_005256329.4 XP_005256386.1 1 - 2 288 96
XM_006721350.2 XP_006721413.1 1 - 2 288 96
XM_011523476.2 XP_011521778.1 1 - 2 288 96
XM_011523479.2 XP_011521781.1 1 - 2 288 96
GenBank mRNA AI458261.1 - 1 - 2 228 96

View Full Product Details