The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004525.2 | NP_004516.2 | 13 - 14 | 1986 | 107 |
XM_011511183.2 | XP_011509485.1 | 13 - 14 | 1987 | 107 | |
GenBank mRNA | AY265357.1 | - | 13 - 14 | 1840 | 107 |
AY265358.1 | - | 13 - 14 | 1837 | 107 | |
U33837.1 | - | 13 - 14 | 1877 | 107 |