The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004525.2 | NP_004516.2 | 50 - 51 | 10057 | 69 |
XM_011511183.2 | XP_011509485.1 | 50 - 51 | 10058 | 69 | |
XM_011511184.2 | XP_011509486.1 | 35 - 36 | 7570 | 69 | |
GenBank mRNA | AY265357.1 | - | 50 - 51 | 9911 | 69 |
AY265358.1 | - | 50 - 51 | 9908 | 69 | |
U04441.1 | - | 8 - 9 | 1735 | 69 | |
U33837.1 | - | 50 - 51 | 9948 | 69 |