The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004525.2 | NP_004516.2 | 52 - 53 | 10376 | 143 |
XM_011511183.2 | XP_011509485.1 | 52 - 53 | 10377 | 143 | |
XM_011511184.2 | XP_011509486.1 | 37 - 38 | 7889 | 143 | |
GenBank mRNA | AK302843.1 | - | 3 - 4 | 366 | 143 |
AY265357.1 | - | 52 - 53 | 10230 | 143 | |
AY265358.1 | - | 52 - 53 | 10227 | 143 | |
U04441.1 | - | 10 - 11 | 2054 | 143 | |
U33837.1 | - | 52 - 53 | 10267 | 143 |