The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003165.3 | NP_003156.1 | 19 - 20 | 2021 | 85 |
GenBank mRNA | AF004563.1 | - | 19 - 20 | 1944 | 85 |
AK295154.1 | - | 18 - 19 | 1879 | 85 | |
AK295802.1 | - | 14 - 15 | 1447 | 85 |