The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022820.4 | NP_073731.1 | 3 - 4 | 317 | 135 |
NM_057095.2 | NP_476436.1 | 3 - 4 | 317 | 135 | |
NM_057096.3 | NP_476437.1 | 3 - 4 | 317 | 135 | |
NR_103869.1 | 3 - 4 | 317 | 135 | ||
GenBank mRNA | AF280108.1 | - | 3 - 4 | 214 | 135 |
AF280109.1 | - | 3 - 4 | 214 | 135 | |
AF280110.1 | - | 3 - 4 | 214 | 135 | |
AF280111.1 | - | 3 - 4 | 214 | 135 | |
AF319634.1 | - | 3 - 4 | 214 | 135 | |
AF337813.1 | - | 3 - 4 | 317 | 135 | |
AY390423.1 | - | 3 - 4 | 213 | 135 | |
AY390424.1 | - | 3 - 4 | 214 | 135 | |
AY390425.1 | - | 3 - 4 | 214 | 135 | |
AY390426.1 | - | 3 - 4 | 213 | 135 |