The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030974.3 | NP_112236.3 | 5 - 6 | 1252 | 141 |
NR_038270.1 | 5 - 6 | 1250 | 141 | ||
XM_017013887.1 | XP_016869376.1 | 5 - 6 | 1280 | 141 | |
XM_017013888.1 | XP_016869377.1 | 5 - 6 | 1280 | 141 | |
GenBank mRNA | AB052764.1 | - | 5 - 6 | 1249 | 141 |
AB052765.1 | - | 4 - 5 | 1044 | 141 | |
AK075518.1 | - | 5 - 6 | 1207 | 141 | |
AL136816.1 | - | 5 - 6 | 976 | 141 | |
BC017230.2 | - | 3 - 4 | 354 | 141 | |
BC025244.1 | - | 5 - 6 | 1252 | 141 | |
BC034028.1 | - | 5 - 6 | 781 | 141 | |
FJ655995.1 | - | 5 - 6 | 1038 | 141 |