The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243565.1 | NP_001230494.1 | 5 - 6 | 572 | 101 |
NM_001319959.1 | NP_001306888.1 | 6 - 7 | 712 | 101 | |
NM_024887.3 | NP_079163.2 | 6 - 7 | 689 | 101 | |
NM_205861.2 | NP_995583.1 | 6 - 7 | 689 | 101 | |
XM_006710912.2 | XP_006710975.1 | 6 - 7 | 748 | 101 | |
XM_006710913.2 | XP_006710976.1 | 6 - 7 | 627 | 101 | |
XM_006710914.2 | XP_006710977.1 | 6 - 7 | 624 | 101 | |
XM_006710916.2 | XP_006710979.1 | 6 - 7 | 738 | 101 | |
XM_006710917.2 | XP_006710980.1 | 6 - 7 | 691 | 101 | |
XM_006710918.2 | XP_006710981.1 | 6 - 7 | 647 | 101 | |
XM_011542183.2 | XP_011540485.1 | 6 - 7 | 724 | 101 | |
XM_011542184.2 | XP_011540486.1 | 6 - 7 | 745 | 101 | |
XM_017002379.1 | XP_016857868.1 | 6 - 7 | 627 | 101 | |
XM_017002380.1 | XP_016857869.1 | 6 - 7 | 724 | 101 | |
XM_017002381.1 | XP_016857870.1 | 6 - 7 | 738 | 101 | |
XM_017002382.1 | XP_016857871.1 | 6 - 7 | 647 | 101 | |
GenBank mRNA | AB090852.1 | - | 5 - 6 | 550 | 101 |
AK023164.1 | - | 6 - 7 | 619 | 101 | |
AK308187.1 | - | 6 - 7 | 631 | 101 | |
BC003643.2 | - | 5 - 6 | 594 | 101 | |
BC004117.2 | - | 5 - 6 | 514 | 101 | |
BX648507.1 | - | 6 - 7 | 714 | 101 | |
GU727641.1 | - | 5 - 6 | 594 | 101 |