Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACTL6B
Entrez Gene ID
51412
Gene Name
actin like 6B
Gene Aliases
ACTL6, BAF53B, arpNalpha
Chromosome Location
Chr.7: 100643097 - 100656461 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016188.4 NP_057272.1 11 - 12 1128 64
NR_134539.1 11 - 12 1128 64
GenBank mRNA AB015906.1 - 11 - 12 1092 64
AF041475.1 - 11 - 12 1091 64
AK314940.1 - 11 - 12 1128 64
BC020944.1 - 11 - 12 1115 64
BM696009.1 - 1 - 2 46 64

View Full Product Details