The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016188.4 | NP_057272.1 | 8 - 9 | 871 | 62 |
NR_134539.1 | 8 - 9 | 871 | 62 | ||
GenBank mRNA | AB015906.1 | - | 8 - 9 | 835 | 62 |
AF041475.1 | - | 8 - 9 | 834 | 62 | |
AK314940.1 | - | 8 - 9 | 871 | 62 | |
BC020944.1 | - | 8 - 9 | 858 | 62 |