Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARFRP1
Entrez Gene ID
10139
Gene Name
ADP ribosylation factor related protein 1
Gene Aliases
ARL18, ARP, Arp1
Chromosome Location
Chr.20: 63698642 - 63708013 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267546.2 NP_001254475.1 6 - 7 566 104
NM_001267547.2 NP_001254476.1 7 - 8 665 104
NM_001267548.2 NP_001254477.1 7 - 8 562 104
NM_003224.5 NP_003215.1 7 - 8 660 104
NR_051954.2 7 - 8 658 104
NR_051955.2 7 - 8 663 104
NR_051956.2 7 - 8 660 104
NR_051957.2 7 - 8 655 104
NR_051958.2 6 - 7 589 104
XM_011528482.2 XP_011526784.1 7 - 8 597 104
XM_011528483.1 XP_011526785.1 7 - 8 727 104
XM_017027576.1 XP_016883065.1 7 - 8 541 104
GenBank mRNA AL533893.3 - 7 - 8 650 104
BC021023.1 - 7 - 8 545 104
BC092480.1 - 7 - 8 546 104
BC093967.1 - 7 - 8 563 104
BC143467.1 - 7 - 8 568 104
BI819763.1 - 7 - 8 532 104
BM546222.1 - - 585 104
BT019624.1 - 6 - 7 512 104
BT019663.1 - 6 - 7 512 104
BX342805.2 - 6 - 7 568 104
BX463457.2 - 7 - 8 562 104
CR456752.1 - 6 - 7 512 104
X91504.1 - 6 - 7 523 104

View Full Product Details