Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PGF
Entrez Gene ID
5228
Gene Name
placental growth factor
Gene Aliases
D12S1900, PGFL, PIGF, PLGF, PlGF-2, SHGC-10760
Chromosome Location
Chr.14: 74941830 - 74955764 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001207012.1 NP_001193941.1 1 - 2 599 113
NM_002632.5 NP_002623.2 1 - 2 599 113
GenBank mRNA A18411.1 - 1 - 2 398 113
AK309886.1 - 1 - 2 432 113
AK310608.1 - 1 - 2 432 113
BC001422.2 - 1 - 2 423 113
BC007255.1 - 1 - 2 404 113
BC007789.2 - 1 - 2 401 113
BT007182.1 - 1 - 2 77 113
BX248289.1 - 1 - 2 599 113
S72960.1 - 1 - 2 89 113
X54936.1 - 1 - 2 398 113

View Full Product Details