Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYVE1
Entrez Gene ID
10894
Gene Name
lymphatic vessel endothelial hyaluronan receptor 1
Gene Aliases
CRSBP-1, HAR, LYVE-1, XLKD1
Chromosome Location
Chr.11: 10557866 - 10568818 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006691.3 NP_006682.2 3 - 4 685 75
GenBank mRNA AF127670.2 - 3 - 4 519 75
AK075443.1 - 3 - 4 599 75
AK312462.1 - 3 - 4 683 75
AY358925.1 - 3 - 4 558 75
BC026231.1 - 3 - 4 444 75

View Full Product Details