Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZMIZ1
Entrez Gene ID
57178
Gene Name
zinc finger MIZ-type containing 1
Gene Aliases
MIZ, RAI17, TRAFIP10, ZIMP10, hZIMP10
Chromosome Location
Chr.10: 79068994 - 79316528 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020338.3 NP_065071.1 19 - 20 2859 80
XM_005269987.4 XP_005270044.1 19 - 20 2987 80
XM_005269988.3 XP_005270045.1 19 - 20 2918 80
XM_006717923.3 XP_006717986.1 19 - 20 2915 80
XM_006717924.3 XP_006717987.1 18 - 19 2837 80
XM_006717925.3 XP_006717988.1 19 - 20 2918 80
XM_011539978.2 XP_011538280.1 13 - 14 2191 80
XM_011539979.1 XP_011538281.1 13 - 14 2317 80
XM_011539980.2 XP_011538282.1 13 - 14 2127 80
XM_017016440.1 XP_016871929.1 13 - 14 3137 80
XM_017016441.1 XP_016871930.1 13 - 14 2095 80
XM_017016442.1 XP_016871931.1 13 - 14 2299 80
XM_017016443.1 XP_016871932.1 13 - 14 2137 80
GenBank mRNA AB033050.2 - 13 - 14 2078 80
AF357909.1 - 14 - 15 1999 80
AY235683.1 - 16 - 17 2356 80

View Full Product Details