The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020338.3 | NP_065071.1 | 24 - 25 | 3670 | 67 |
XM_005269987.4 | XP_005270044.1 | 24 - 25 | 3798 | 67 | |
XM_005269988.3 | XP_005270045.1 | 24 - 25 | 3729 | 67 | |
XM_006717923.3 | XP_006717986.1 | 24 - 25 | 3726 | 67 | |
XM_006717924.3 | XP_006717987.1 | 23 - 24 | 3648 | 67 | |
XM_011539978.2 | XP_011538280.1 | 18 - 19 | 3002 | 67 | |
XM_011539979.1 | XP_011538281.1 | 18 - 19 | 3128 | 67 | |
XM_011539980.2 | XP_011538282.1 | 18 - 19 | 2938 | 67 | |
XM_017016440.1 | XP_016871929.1 | 18 - 19 | 3948 | 67 | |
XM_017016441.1 | XP_016871930.1 | 18 - 19 | 2906 | 67 | |
XM_017016442.1 | XP_016871931.1 | 18 - 19 | 3110 | 67 | |
XM_017016443.1 | XP_016871932.1 | 18 - 19 | 2948 | 67 | |
GenBank mRNA | AB033050.2 | - | 18 - 19 | 2889 | 67 |
AF357909.1 | - | 20 - 21 | 2801 | 67 | |
AY235683.1 | - | 21 - 22 | 3167 | 67 | |
BC025342.1 | - | 1 - 2 | 233 | 67 | |
BC035890.1 | - | 1 - 2 | 47 | 67 |