The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143976.1 | NP_001137448.1 | 6 - 7 | 1390 | 93 |
NM_003390.3 | NP_003381.1 | 6 - 7 | 1530 | 93 | |
GenBank mRNA | AK122837.1 | - | 6 - 7 | 1390 | 93 |
BC051831.1 | - | 6 - 7 | 1328 | 93 | |
BC070052.1 | - | 6 - 7 | 1520 | 93 | |
U10564.1 | - | 6 - 7 | 1530 | 93 | |
X62048.1 | - | 6 - 7 | 1447 | 93 |