The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014495.3 | NP_055310.1 | 3 - 4 | 805 | 112 |
GenBank mRNA | AF152562.1 | - | 3 - 4 | 749 | 112 |
AK289426.1 | - | 3 - 4 | 772 | 112 | |
AK315304.1 | - | 3 - 4 | 775 | 112 | |
AY358273.1 | - | 3 - 4 | 761 | 112 | |
BC007059.1 | - | 3 - 4 | 772 | 112 | |
BC058287.1 | - | 3 - 4 | 776 | 112 |