The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014495.3 | NP_055310.1 | 4 - 5 | 913 | 113 |
GenBank mRNA | AF152562.1 | - | 4 - 5 | 857 | 113 |
AK315304.1 | - | 4 - 5 | 883 | 113 | |
AY358273.1 | - | 4 - 5 | 869 | 113 | |
BC007059.1 | - | 4 - 5 | 880 | 113 | |
BC058287.1 | - | 4 - 5 | 884 | 113 |