Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP13A2
Entrez Gene ID
23400
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
Chromosome Location
Chr.1: 16985958 - 17011972 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001141973.2 NP_001135445.1 24 - 25 2981 111
NM_001141974.2 NP_001135446.1 23 - 24 2864 111
NM_022089.3 NP_071372.1 24 - 25 2996 111
XM_005245810.1 XP_005245867.1 24 - 25 2993 111
XM_005245811.1 XP_005245868.1 24 - 25 2981 111
XM_005245812.1 XP_005245869.1 24 - 25 2969 111
XM_005245815.1 XP_005245872.1 23 - 24 2879 111
XM_006710512.1 XP_006710575.1 24 - 25 2978 111
XM_006710513.1 XP_006710576.1 24 - 25 2954 111
XM_011541128.1 XP_011539430.1 24 - 25 2981 111
XM_011541129.1 XP_011539431.1 23 - 24 2789 111
XM_017000844.1 XP_016856333.1 24 - 25 2981 111
XM_017000845.1 XP_016856334.1 24 - 25 2978 111
XM_017000846.1 XP_016856335.1 24 - 25 2954 111
XM_017000847.1 XP_016856336.1 24 - 25 2951 111
XM_017000848.1 XP_016856337.1 23 - 24 2879 111
XM_017000849.1 XP_016856338.1 23 - 24 2864 111
XM_017000850.1 XP_016856339.1 23 - 24 2789 111
GenBank mRNA AJ009947.1 - 2 - 3 202 111
AK025928.1 - 4 - 5 385 111
AK075310.1 - 14 - 15 1778 111
AK290210.1 - 24 - 25 2937 111
AL354615.1 - 24 - 25 2796 111
AL833966.1 - 5 - 6 650 111
AY461712.1 - 24 - 25 2795 111
BC030267.1 - 23 - 24 2807 111
BC034575.1 - 21 - 22 2487 111

View Full Product Details