Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP13A2
Entrez Gene ID
23400
Gene Name
ATPase 13A2
Gene Aliases
CLN12, HSA9947, KRPPD, PARK9
Chromosome Location
Chr.1: 16985958 - 17011972 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001141973.2 NP_001135445.1 7 - 8 852 109
NM_001141974.2 NP_001135446.1 7 - 8 852 109
NM_022089.3 NP_071372.1 7 - 8 867 109
XM_005245810.1 XP_005245867.1 7 - 8 864 109
XM_005245811.1 XP_005245868.1 7 - 8 852 109
XM_005245812.1 XP_005245869.1 7 - 8 867 109
XM_005245815.1 XP_005245872.1 7 - 8 867 109
XM_006710512.1 XP_006710575.1 7 - 8 849 109
XM_006710513.1 XP_006710576.1 7 - 8 852 109
XM_011541128.1 XP_011539430.1 7 - 8 867 109
XM_011541129.1 XP_011539431.1 7 - 8 867 109
XM_017000844.1 XP_016856333.1 7 - 8 867 109
XM_017000845.1 XP_016856334.1 7 - 8 849 109
XM_017000846.1 XP_016856335.1 7 - 8 852 109
XM_017000847.1 XP_016856336.1 7 - 8 864 109
XM_017000848.1 XP_016856337.1 7 - 8 867 109
XM_017000849.1 XP_016856338.1 7 - 8 852 109
XM_017000850.1 XP_016856339.1 7 - 8 867 109
GenBank mRNA AK290210.1 - 7 - 8 808 109
AL354615.1 - 7 - 8 667 109
AY461712.1 - 7 - 8 666 109
BC030267.1 - 7 - 8 795 109
BC034575.1 - 4 - 5 358 109

View Full Product Details