The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019617.3 | NP_062563.3 | 2 - 3 | 174 | 126 |
GenBank mRNA | AB039886.1 | - | 2 - 3 | 126 | |
AJ966789.1 | - | 2 - 3 | 174 | 126 | |
AY139184.1 | - | 2 - 3 | 112 | 126 | |
AY359050.1 | - | 2 - 3 | 98 | 126 | |
AY456961.1 | - | 2 - 3 | 126 | ||
BC059778.1 | - | 2 - 3 | 100 | 126 |