The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019617.3 | NP_062563.3 | 4 - 5 | 418 | 117 |
GenBank mRNA | AB039886.1 | - | 4 - 5 | 418 | 117 |
AJ966789.1 | - | 4 - 5 | 418 | 117 | |
AY139184.1 | - | 4 - 5 | 356 | 117 | |
AY359050.1 | - | 4 - 5 | 342 | 117 | |
AY456961.1 | - | 4 - 5 | 355 | 117 | |
BC059778.1 | - | 4 - 5 | 344 | 117 |